Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoguanamine, 99%

Catalog No. AAB201570E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB201570E 2500 g
AAB2015722 100 g
AAB2015736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB201570E Supplier Thermo Scientific Chemicals Supplier No. B201570E
Add to Cart

CAS: 91-76-9 | C9H9N5 | 187.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-76-9
Molecular Formula C9H9N5
Molecular Weight (g/mol) 187.21
MDL Number MFCD00023187
InChI Key GZVHEAJQGPRDLQ-UHFFFAOYSA-N
Synonym benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl
PubChem CID 7064
IUPAC Name 6-phenyl-1,3,5-triazine-2,4-diamine
SMILES NC1=NC(=NC(N)=N1)C1=CC=CC=C1

Specifications

Melting Point 225°C to 228°C
Quantity 2500 g
Beilstein 153223
Merck Index 14,1089
Formula Weight 187.21
Percent Purity 99%
Chemical Name or Material Benzoguanamine
Hazard Category H302+H332
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 202-095-6
RTECSNumber XY7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.