Learn More
Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 99.5 |
| CAS Max % | 100.0 |
| Melting Point | 121.0°C to 123.0°C |
| Color | White |
| Boiling Point | 249.0°C |
| Flash Point | 121°C |
| Ignition Residue | 0.005% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.6% |
| Packaging | Plastic drum |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. Causes damage to organs through prolonged or repeated exposure if inhaled. |
| Precautionary Statement | GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for s |
| EINECSNumber | 200-618-2 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.