Learn More
Apexbio Technology LLC Bisphenol B 77-40-7 10g
Supplier: Apexbio Technology LLC A992710000
Bisphenol B (CAS 77-40-7) is a synthetic bisphenol compound structurally analogous to bisphenol A It functions as an endocrine-disrupting chemical by binding to and activating estrogen receptors thereby mimicking the action of endogenous estrogens and potentially altering hormonal signaling pathways Owing to these properties Bisphenol B is widely utilized in toxicological and biomedical research to investigate endocrine disruption reproductive toxicity and possible health risks associated with environmental exposure to bisphenol analogues Its study supports risk assessment and informs regulatory evaluations of bisphenol-related substances
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.