Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone Dimethylketal 98.0+%, TCI America™
SDP

Catalog No. B199925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B199925G 25 g
B19995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B199925G Supplier TCI America Supplier No. B199925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2235-01-0
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00025900
InChI Key NYRVXYOKUZSUDA-UHFFFAOYSA-N
Synonym Dimethoxydiphenylmethane
PubChem CID 75228
IUPAC Name [dimethoxy(phenyl)methyl]benzene
SMILES COC(OC)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 106°C
Color White-Yellow
Boiling Point 290°C
Quantity 25 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzophenone Dimethylketal
EINECSNumber (4)-1560
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.