Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone hydrazone, 98+%

Catalog No. AC105571000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105571000 100 g Glass bottle
AC105570050 5 g Glass bottle
AC105575000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC105571000 Supplier Thermo Scientific Chemicals Supplier No. 105571000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 5350-57-2 | C13H12N2 | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5350-57-2
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.25
MDL Number MFCD00007624
InChI Key QYCSNMDOZNUZIT-UHFFFAOYSA-N
Synonym benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
PubChem CID 79304
IUPAC Name benzhydrylidenehydrazine
SMILES C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 95°C to 99°C
Color White to Yellow
Boiling Point 225°C to 230°C (55.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (C6H5)2C=NNH2
Quantity 100 g
Beilstein 07, 417
Fieser 13,242
Formula Weight 196.25
Percent Purity 98+%
Physical Form Crystalline Powder or Crystalline Needles
Chemical Name or Material Benzophenone hydrazone
Show More Show Less
EINECSNumber 226-321-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.