Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Benzopyrene Monoclonal Antibody (BAP-13)
GREENER_CHOICE

Catalog No. 01676029
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
01-676-029 100 μg
1 options

Catalog No. 01-676-029

Supplier: Invitrogen™ MA119273

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody The antibody BAP-13 is specific for Polynuclear Aromatic Hydrocarbon and inhibits DNA adduct formation (this ability was investigated in rat liver microsomes spiked with calf thymus DNA and 7,8-diol-B[a]P).

Benzo[a]pyrene, also known as 3, 4-Benzopyrene or 3, 4-BP, is classified as a member of the Benzopyrenes. Benzopyrenes are organic compounds containing a benzene fused to a pyrene(benzo[def]phenanthrene) ring system. Benzo[a]pyrene is formally rated as a carcinogenic (IARC 1) potentially toxic compound. 3 4-Benzpyrene is a crystalline, aromatic hydrocarbon consisting of five fused benzene rings and formed during the incomplete combustion of organic matter. 3, 4-Benzpyrene is primarily found in gasoline and diesel exhaust, cigarette smoke, coal tar and coal tar pitch, charcoal-broiled foods and certain other foods, amino acids, fatty acids and carbohydrate pyrolysis products, soot smoke, creosote oil, petroleum asphalt and shale oils. This substance is used only for research purposes. 3, 4-Benzpyrene is reasonably anticipated to be a human carcinogen. [National Center for Biotechnology Information. PubChem Compound Database; CID=2336].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Benzopyrene
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone BAP-13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Alias 4-Benzopyrene; 4-BP; Benzo(a)pyrene; C20H12
Host Species Mouse
Immunogen Benzo[a]pyrenyl-1-butyric acid conjugated to BSA..
Purification Method Sequential Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.