Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl Bromide 98.0+%, TCI America™
SDP

Catalog No. B0104500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0104500G 500 g
B010425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0104500G Supplier TCI America Supplier No. B0104500G

Chemical Identifiers

CAS 618-32-6
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00000112
InChI Key AQIHMSVIAGNIDM-UHFFFAOYSA-N
Synonym benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide
PubChem CID 12056
IUPAC Name benzoyl bromide
SMILES BrC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 219°C
Quantity 500 g
UN Number 3265
Formula Weight 185.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzoyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.