Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl Cyanide 98%, Thermo Scientific™

Catalog No. AC152060250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152060250 25 g
AC152061000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152060250 Supplier Thermo Scientific Chemicals Supplier No. 152060250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-90-1
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.134
InChI Key GJQBHOAJJGIPRH-UHFFFAOYSA-N
Synonym phenylglyoxylonitrile, benzoyl nitrile, benzoylcyanide, oxo phenyl acetonitrile, alpha-oxobenzeneacetonitrile, benzeneacetonitrile, .alpha.-oxo, glyoxylonitrile, phenyl, alpha-oxo-alpha-tolunitrile, 2-oxo-2-phenylethanenitrile, alpha-tolunitrile, alpha-oxo
PubChem CID 11953
ChEBI CHEBI:51853
IUPAC Name benzoyl cyanide
SMILES C1=CC=C(C=C1)C(=O)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29°C to 32°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Linear Formula C6H5COCN
Quantity 25 g
Beilstein 10, 659
Fieser 06,35; 13,54
Formula Weight 131.13
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material Benzoyl cyanide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 210-359-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.