Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. B35675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35675G Supplier TCI America Supplier No. B35675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A mild reagent for benzoylation.

Chemical Identifiers

CAS 36967-85-8
Molecular Formula C8H5F3O4S
Molecular Weight (g/mol) 254.18
MDL Number MFCD17013536
InChI Key GEZVJNXOBFDIPM-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid Benzoyl Ester, Benzoyl Triflate
PubChem CID 11064956
IUPAC Name trifluoromethanesulfonyl benzoate
SMILES FC(F)(F)S(=O)(=O)OC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 254.18
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material Benzoyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.