Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Benzoylecgonine Monoclonal Antibody (1P3G2)
GREENER_CHOICE

Catalog No. PIMA514691 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514691 Supplier Invitrogen™ Supplier No. MA514691
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA5-14691 targets Benzoylecgonine in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14691 immunogen is benzoylecgonine-KLH. Product MA514691 is a smaller package size of MIB0202 (formerly sold as a Seradyn product).

Benzoylecgonine is used as the main pharmaceutical ingredient in the prescription drug Esterom, a topical solution used for the relief of muscle pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Benzoylecgonine
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1P3G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene Alias C16H19NO4
Host Species Mouse
Immunogen Benzoylecgonine-KLH.
Purification Method Ion-exchange Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.