Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylferrocene 98.0+%, TCI America™
SDP

Catalog No. B088625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B088625G 25 g
B08865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B088625G Supplier TCI America Supplier No. B088625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1272-44-2
Molecular Formula C17H14FeO
Molecular Weight (g/mol) 290.14
MDL Number MFCD00001431
InChI Key JJHHJZCYDPLHIL-UHFFFAOYNA-N
IUPAC Name Benzoylferrocene
SMILES [Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1

Specifications

Melting Point 110°C
Quantity 25 g
Formula Weight 290.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.