Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylurea 98.0+%, TCI America™
SDP

Catalog No. B0400500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0400500G 500 g
B040025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0400500G Supplier TCI America Supplier No. B0400500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 614-22-2
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00025441
InChI Key HRYILSDLIGTCOP-UHFFFAOYSA-N
Synonym benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide
PubChem CID 69190
ChEBI CHEBI:39416
IUPAC Name benzoylurea
SMILES NC(=O)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 210°C
Color White
Quantity 500 g
Formula Weight 164.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylurea
RTECSNumber YR9070000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.