Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 2,2,2-Trifluoro-N-phenylacetimidate 98.0+%, TCI America™
SDP

Catalog No. B32341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32341G 1 g
B32345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32341G Supplier TCI America Supplier No. B32341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 952057-61-3
Molecular Formula C15H12F3NO
Molecular Weight (g/mol) 279.262
InChI Key KVGZVQNJUZHPJJ-UHFFFAOYSA-N
Synonym 2,2,2-Trifluoro-N-phenylacetimidic Acid Benzyl Ester
PubChem CID 23583338
IUPAC Name benzyl 2,2,2-trifluoro-N-phenylethanimidate
SMILES C1=CC=C(C=C1)COC(=NC2=CC=CC=C2)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 279.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 2,2,2-Trifluoro-N-phenylacetimidate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.