Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™
SDP

Catalog No. B4627100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4627100G 100 g
B462725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4627100G Supplier TCI America Supplier No. B4627100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2495-35-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00048147
InChI Key GCTPMLUUWLLESL-UHFFFAOYSA-N
Synonym benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
PubChem CID 75617
IUPAC Name benzyl prop-2-enoate
SMILES C=CC(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 60°C
Quantity 100 g
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Acrylate (stabilized with MEHQ)
EINECSNumber (3)-2873
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.