Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, ACS Reagent, ≥99%, Solstice

Catalog No. 60046790 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-790 500 mL Glass Bottle
60-046-787 100 mL Glass Bottle
60-046-788 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-790 Supplier Solstice Supplier No. 402834500ML

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -16°C to -13°C
Density 1.045g/mL at 25°C
Boiling Point 203°C to 205°C
Flash Point 96°C (204.8°F)
Ignition Residue ≤0.005%
Ignition Point 817°F
Packaging Glass Bottle
Refractive Index n20/D 1.539
Linear Formula C6H5CH2OH
Quantity 500 mL
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Vapor Pressure 13.3mmHg (100°C), 3.75mmHg (77°C)
Formula Weight 108.14g/mol
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Benzyl alcohol
Show More Show Less
Hazard Statement P261-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338
DOTInformation UN 3334
RTECSNumber 202-859-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.