Learn More
Benzyl Alcohol, ACS Reagent, Reagents
Specifications
Specifications
| Boiling Range | 205°C |
| Melting Point | -15.4°C |
| Color | Colorless |
| pH | Neutral |
| Boiling Point | 205°C |
| Melting Range | -15.4°C |
| Packaging | Glass Bottle |
| Quantity | 4 L |
| Specific Gravity | 1.044 |
| Grade | ACS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.