Learn More
Description
Benzyl Benzoate USP is clear, colorless, oily liquid having a slight aromatic odor.
Alternative name(s): Benzoic acid, benzoic ester, phenylmethyl ester
The freezing point of BENZYL BENZOATE USP is 18 - 21 °C (64 - 70 °F). It will begin to liquefy at 18 °C (64°F).
Product is not affected by cycling between freezing and thawing conditions as long as label directions are followed.
Product can be left at room temperature or in a controlled-temperature water bath or a hot box set to 50°C (122°F) to thaw. Gently stir or tumble the container to ensure it is homogenous.
It is recommended to use the minimum amount of heat necessary to thaw the product. It is also advised not to perform direct heating to higher temperatures.
Specifications
Specifications
| Chemical Name or Material | Benzyl Benzoate, USP |
| Packaging | Jug, 4L, Amber Glass, Round, 38-439 (for MCP38439LNC) |
| CAS | 120-51-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 212.2468 |
| Quantity | 4 L |
| Percent Purity | 99 to 100.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
