Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Benzyl Benzoate, USP
SDP

Catalog No. 14186063 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-063 500 mL
14-186-060 4 L
14-185-591 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-186-063 Supplier Medisca Supplier No. 103008
Add to Cart
Add to Cart

Benzyl Benzoate USP is clear, colorless, oily liquid having a slight aromatic odor.

Benzyl Benzoate USP is clear, colorless, oily liquid having a slight aromatic odor.

Alternative name(s): Benzoic acid, benzoic ester, phenylmethyl ester

The freezing point of BENZYL BENZOATE USP is 18 - 21 °C (64 - 70 °F). It will begin to liquefy at 18 °C (64°F).

Product is not affected by cycling between freezing and thawing conditions as long as label directions are followed.

Product can be left at room temperature or in a controlled-temperature water bath or a hot box set to 50°C (122°F) to thaw. Gently stir or tumble the container to ensure it is homogenous.

It is recommended to use the minimum amount of heat necessary to thaw the product. It is also advised not to perform direct heating to higher temperatures.

Specifications

Chemical Name or Material Benzyl Benzoate, USP
Packaging Bottle, Amber Glass, Boston Round, 500mL, 38-439
CAS 120-51-4
Type Drug Standard
Molecular Weight (g/mol) 212.2468
Quantity 500 mL
Percent Purity 99 to 100.5 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.