Learn More
Benzyl methacrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol
Description
Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light and heat. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.04 |
| Boiling Point | 231°C to 233°C |
| Flash Point | 107°C (225°F) |
| Refractive Index | 1.514 |
| Quantity | 1000 g |
| Beilstein | 2045924 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 176.22 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 219-674-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.