Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzylamine

Catalog No. 407712800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-771-2800ML 800 mL
40-771-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-771-2800ML Supplier Sigma Aldrich Supplier No. 407712800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzylamine

Specifications

CAS 100-46-9
Melting Point 10°C (lit.)
Density 0.981 g/mL (at 25°C (literature))
Boiling Point 184°C to 185°C (lit.)
Molecular Formula C7H9N
Refractive Index n20/D 1.543 (literature)
Linear Formula C6H5CH2NH2
MDL Number MFCD00008106
Quantity 800 mL
Synonym alpha-Aminotoluene
Molecular Weight (g/mol) 107.15
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.