Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B0407100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0407100G 100 g
B040725G 25 g
B0407500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0407100G Supplier TCI America Supplier No. B0407100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3287-99-8
Molecular Formula C7H10ClN
Molecular Weight (g/mol) 143.614
MDL Number MFCD00012852
InChI Key XKXHCNPAFAXVRZ-UHFFFAOYSA-N
Synonym benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
PubChem CID 2724127
IUPAC Name phenylmethanamine;hydrochloride
SMILES C1=CC=C(C=C1)CN.Cl

Specifications

Melting Point 253°C
Color White
Quantity 100 g
Formula Weight 143.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylamine Hydrochloride
EINECSNumber (3)-0367
RTECSNumber DP5425000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.