Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyldimethylstearylammonium Chloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. B129725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B129725G 25 g
B1297100G 100 g
B1297500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B129725G Supplier TCI America Supplier No. B129725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122-19-0
Molecular Formula C27H50ClN
Molecular Weight (g/mol) 424.154
MDL Number MFCD00011776
InChI Key SFVFIFLLYFPGHH-UHFFFAOYSA-M
Synonym Octadecyldimethylbenzylammonium Chloride, Benzyldimethyloctadecylammonium Chloride
PubChem CID 31204
IUPAC Name benzyl-dimethyl-octadecylazanium;chloride
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]

Specifications

Color White
Quantity 25 g
Formula Weight 424.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethylstearylammonium Chloride Hydrate
EINECSNumber (3)-0326&(3)-2694
RTECSNumber BO7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.