Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylmalonic Acid 98.0+%, TCI America™
SDP

Catalog No. B042225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B042225G 25 g
B04225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B042225G Supplier TCI America Supplier No. B042225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-75-1
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004261
InChI Key JAEJSNFTJMYIEF-UHFFFAOYSA-N
PubChem CID 12031
IUPAC Name 2-benzylpropanedioic acid
SMILES OC(=O)C(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 118°C
Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylmalonic Acid
RTECSNumber OO0525000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.