Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylsulfonamide 98.0+%, TCI America™
SDP

Catalog No. B34665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34665G Supplier TCI America Supplier No. B34665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4563-33-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00041893
InChI Key ABOYDMHGKWRPFD-UHFFFAOYSA-N
Synonym benzenemethanesulfonamide, benzylsulfonamide, alpha-toluenesulfonamide, methanesulfonamide, 1-phenyl, .alpha.-toluenesulfonamide, 1-phenylmethanesulfonamide, toluene-.omega.-sulfonamide, toluene-alpha-sulphonamide, benzyl-sulfonamide, a-toluenesulfonamide
PubChem CID 78318
IUPAC Name phenylmethanesulfonamide
SMILES NS(=O)(=O)CC1=CC=CC=C1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzylsulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.