Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriethoxysilane 96.0+%, TCI America™
SDP

Catalog No. B328225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B328225G 25 g
B32821G 1 g
B32825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B328225G Supplier TCI America Supplier No. B328225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2549-99-7
Molecular Formula C13H22O3Si
Molecular Weight (g/mol) 254.40
MDL Number MFCD00026751
InChI Key CPLASELWOOUNGW-UHFFFAOYSA-N
PubChem CID 75690
IUPAC Name benzyltriethoxysilane
SMILES CCO[Si](CC1=CC=CC=C1)(OCC)OCC

Specifications

Color Yellow
Boiling Point 248°C
Quantity 25 g
Formula Weight 254.40
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyltriethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.