Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltrimethylammonium Chloride 99.0+%, TCI America™
SDP

Catalog No. B0447100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0447100G 100 g
B044725G 25 g
B0447500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0447100G Supplier TCI America Supplier No. B0447100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-93-9
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00011782
InChI Key KXHPPCXNWTUNSB-UHFFFAOYSA-M
Synonym benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
PubChem CID 5963
IUPAC Name benzyltrimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 239°C
Color White
pH 5.0 - 7.0 (50g/L)
Quantity 100 g
UN Number 2811
Formula Weight 185.70
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Chloride
EINECSNumber (3)-2694
RTECSNumber BO8400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.