Learn More
Medchemexpress LLC Benzyltrimethylammonium tribromide | 111865-47-5 | MFCD00134423 | 98.0% | 389.95 g/mol | C10H16Br3N | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01845450G
Encompass_Preferred
Benzyltrimethylammonium tribromide is a quaternary ammonium brominating reagent for research use. It provides controlled bromide equivalents for selective bromination, cyclization, and oxidation reactions, and is supplied as a crystalline powder with high purity and storage recommendations.
  • Provides controlled bromide for selective bromination.
  • Enables cyclization and oxidation reactions with reduced side reactions.
  • High purity for reproducible, analytical-grade performance.
  • Available in multiple laboratory-scale sizes for flexible use.
  • Stable as a sealed crystalline powder when stored at 4°C.
  • In-solution storage guidance: -80°C (6 months), -20°C (1 month).
Catalog No. 50-003-01336