Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltrimethylsilane 96.0+%, TCI America™
SDP

Catalog No. B185625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B185625ML 25 mL
B18565ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B185625ML Supplier TCI America Supplier No. B185625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 770-09-2
Molecular Formula C10H16Si
Molecular Weight (g/mol) 164.323
MDL Number MFCD00039796
InChI Key MRIWRLGWLMRJIW-UHFFFAOYSA-N
Synonym trimethylbenzylsilane, silane, trimethyl phenylmethyl, silane, benzyltrimethyl, alpha-trimethylsilyltoluene, benzyl trimethyl silane, unii-hfx8b6q8wc, hfx8b6q8wc, benzene, trimethylsilyl methyl, amtsi018
PubChem CID 69862
IUPAC Name benzyl(trimethyl)silane
SMILES C[Si](C)(C)CC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 191°C
Quantity 25 mL
UN Number 1993
Formula Weight 164.32
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyltrimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.