Learn More
Medchemexpress LLC Beperidium iodide | 86434-57-3 | 100.0% | 527.44 g·mol⁻¹ | C23H34IN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10015210MG
Encompass_Preferred
Beperidium iodide is a research-grade muscarinic acetylcholine receptor antagonist reported with a pA2 of 7.93. Supplied as the iodide salt, it is intended for laboratory research use only and provides a defined, high-purity material for receptor pharmacology studies.
  • High purity suitable for research applications (reported 99.97%).
  • Identified by CAS 86434-57-3 for unambiguous chemical identification.
  • Chemical formula C23H34IN3O3 and molecular weight 527.44 g·mol⁻¹.
  • Available in small mass quantities for assay development and screening.
  • Provided as the iodide salt optimized for receptor-binding studies.
Catalog No. 50-002-15733