Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berberine Chloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. B045025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B045025G 25 g
B04505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B045025G Supplier TCI America Supplier No. B045025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 141433-60-5
Molecular Formula C20H18ClNO4
Molecular Weight (g/mol) 371.82
MDL Number MFCD00011939
InChI Key VKJGBAJNNALVAV-UHFFFAOYSA-M
Synonym berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
PubChem CID 155074
IUPAC Name 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-13-ylium chloride
SMILES [Cl-].COC1=C(OC)C2=C[N+]3=C(C=C2C=C1)C1=CC2=C(OCO2)C=C1CC3

Specifications

Melting Point 145°C
Quantity 25 g
Formula Weight 371.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Powder
Chemical Name or Material Berberine Chloride Hydrate
RTECSNumber DR9866400
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.