Learn More
Indofine Chemical Bergaptol, 98%, 100 Mg
Supplier: Indofine Chemical 19230 100 MG
BERGAPTOL 100 MG, Coumarin, 4-Hydroxypsoralen; 5-Hydroxypsoralen, C11H6O4, MW: 202.17, 98%, 486-60-2, MFCD00210479
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.