Learn More
Siemens Healthineers Berichrom α2-Antiplasmin Reagent
Catalog No. 23044692 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 5 mL
Catalog No. Quantity
23-044-692 3 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-692 Supplier Siemens Healthineers Supplier No. 10446427
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantative determination of α2-antiplasmin activity as an aid in the diagnosis of inherited or acquired deficiencies and in management of fibrinolytic therapy.
Berichrom α2-Antiplasmin Reagent is used for the determination of α2-antiplasmin and the detection of hereditary or acquired α2-antiplasmin deficiencies. The chromogenic activity assay is also applicable for the monitoring of substitution therapy.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Fibrinolysis Testing
Quantity 3 x 5 mL