Learn More
Siemens Healthineers Berichrom Plasminogen Reagent
Catalog No. 23044693 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 5 mL
Catalog No. Quantity
23-044-693 3 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-693 Supplier Siemens Healthineers Supplier No. 10446431
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative determination of plasminogen activity as an aid in the diagnosis and monitoring of fibrinolytic disorders.
Berichrom Plasminogen, a chromogenic activity assay, is used for the determination of plasminogen, detection of hereditary or acquired plasminogen deficiencies, and the monitoring of fibrinolytic therapy.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Fibrinolysis Testing
Quantity 3 x 5 mL