Learn More
Siemens Healthineers Berichrom Protein C Reagent
Catalog No. 23044696 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
4 x 5 mL
3 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-696 3 x 10 mL
23-044-697 4 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-696 Supplier Siemens Healthineers Supplier No. 10446499
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantative determination of protein C activity as an aid in the diagnosis of inherited and acquired deficiencies.
Berichrom Protein C, a chromogenic functional activity assay, is used for the detection of hereditary or acquired protein C deficiencies in conjunction with other methods (antigenic determination, protein C clotting method) for the differential diagnosis of different protein C deficiency states. The assay is also used for the monitoring of substitution therapy with protein C concentrates in congenital protein C deficiency. The Berichrom Protein C assay is less susceptible to interfering substances than a clotting assay.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Thrombophilia Testing
Quantity 3 x 10 mL