Learn More
Berkshire™ 360 Isolator Cleaning System
Catalog No. 17987362 Shop All Berkshire Corporation Products
Change view
Click to view available options
Product Type:
Isolater Cleaning Tool
Isolator Cleaning Tool Cover
Material:
Knitted Polyester Elastic
Polyester Nonwoven Elastic, Cellulose
Stainless Steel, Aluminum
Quantity:
1 Ea.
10/Pk.
6/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material Quantity
17-987-362 Isolator Cleaning Tool Cover Knitted Polyester Elastic 10/Pk.
17-987-360 Isolater Cleaning Tool Stainless Steel, Aluminum 1 Ea.
17-987-361 Isolator Cleaning Tool Cover Knitted Polyester Elastic 6/Pk.
17-987-363 Isolator Cleaning Tool Cover Polyester Nonwoven Elastic, Cellulose 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-987-362 Supplier Berkshire™ Supplier No. EC360ECST20
Add to Cart
Add to Cart
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
EasyClean 360 ICT Sterile Knitted Polyester Elastic and Padded Covers (17-987-362)
  • Made from Berkshire's SuperPolx™ 1200A 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior giver, particle and residue performance
  • Non-shedding and low-linting cover per USP requirements
  • Stretch fabric and elastic edge for easy on/off with secure fit on mop head
  • Absorbent polyester knit is abrasion resistant; handles a wide range of cleaning solutions
  • Compatible for use in ISO Class 3/EU Grade A Environments
  • Sterile
Specifications
Product Type Isolator Cleaning Tool Cover
Quantity 10/Pk.
Includes 10 sterile knitted polyester elastic covers and 1 sterile knitted polyester padded cover/pack; 20 packs/case
For Use With (Equipment) EasyClean™ 360 Isolator Cleaning Tool
Description Mop Covers
For Use With (Application) Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Certifications/Compliance ISO Class 3/EU Grade A
Material Knitted Polyester Elastic