Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Heavy Duty Nitrile Coated General Purpose Work Gloves

Catalog No. 189994496 Shop All Showa Best Glove Products
Change view
Click to view available options
Coated Area:
Palm, Fingers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coated Area
18-999-4496 Palm, Fingers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4496 Supplier SHOWA™ Supplier No. 391010
This item is not returnable. View return policy

Designed for longevity, these heavy-duty general purpose work gloves feature a latex palm with an additional wrinkle-finished nitrile coating.

  • Heavy-duty general purpose work gloves
  • Latex palm with an additional wrinkle-finished nitrile coating
  • Soft cotton jersey provides comfort and flexibility
  • Reinforced safety cuff

Specifications

Coated Area Palm, Fingers
Construction Jersey Knit
Coating Material Nitrile
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Construction, Glass Handling, Manufacturing, and Packaging
Glove Cuff Color Natural
Glove Cuff Style Safety
Grip Design Crinkle Finish
Liner Material Cotton Jersey
Series 3910
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.