SHOWA™ Black Knight™ PVC-Coated Gauntlets

Durable and economical

Manufacturer: SHOWA™ 771010

 View more versions of this product

Catalog No. 11-395-14

  Check Availability
Add to Cart

    Specifications

    Certifications CFIA
    Exterior Finish Smooth
    Glove Cuff Style Gauntlet
    For Use With (Application) any general purpose industrial setting or application
    Length (English) 10 in.
    Length (Metric) 25.4cm
    Lining Jersey
    Material PVC
    Size Large
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit