Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 7703R PVC Coated General Purpose Gloves, Cotton Liner and Knit Wrist

Catalog No. 19019320 Shop All Showa Best Glove Products
Change view
Click to view available options
Rough Finish; Knitwrist
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-019-320 Rough Finish; Knitwrist
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-019-320 Supplier SHOWA™ Supplier No. 7703R10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PVC coated glove provides effective defense against chemicals, oils and grease. Designed for a range of industrial applications.

Ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction

  • Fully coated PVC with rough finish
  • Cotton jersey lined Sanitized™
  • Recyclable
  • Washable

Specifications

Certifications/Compliance CFIA
Color Black
Latex Free Yes
Material PVC
Size Large
Thickness (English) 30 mil
Thickness (Metric) 0.76 mm
For Use With (Application) Automotive repairs and maintenance, maintenance, painting and spray, shipping and receiving
Glove Cuff Style Elasticated
Grip Design Rough
Liner Material Cotton Jersey
Lining Lined
Model 7703R-10
Series 7703R Series
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.