Learn More
SHOWA™ Nitri-Dex™ Flock-Lined Nitrile Gloves
Catalog No. 191501290 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-1290 X-Small
19-150-1291 Small
19-150-1293 Large
19-150-1294 X-Large
19-150-1295 2X-Large
19-150-1292 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-150-1290 Supplier SHOWA™ Supplier No. 707FL06
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unsupported, flock-lined nitrile coated gloves feature a lasting grip and provide protection from oils, hydrocarbons, grease and abrasion.
  • Ideal for jobs requiring comfort, tactility, and contact with chemical hazards
  • Flocked cotton liner ensures easy donning and doffing
  • Rolled cuff prevents debris from entering
  • Lightweight nitrile material helps reduce hand fatigue
Specifications
ANSI Abrasion Rating ANSI 1
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE
Chlorinated No
Color Blue
Latex Free Yes
Material Nitrile
Size X-Small
Thickness (English) 9 mil
Thickness (Metric) 0.23 mm
EN388 Standard EN 388:2003 1001
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Manufacturing, laboratories, paint applications, fuel handling, food processing, janitorial, agricultural chemicals, refineries, light duty maintenance and clean up, paint applications, poultry processing
Glove Cuff Style Gauntlet
Grip Design Tractor tread
Liner Material Cotton Flock
Lining Lined
Model 707FL-06
Series SHOWA™ 707FL
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.