Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 68NFW Wrinkle Latex Coated Aramid Fiber Gloves

Catalog No. 19055098 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-055-098 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-055-098 Supplier SHOWA™ Supplier No. 68NFW10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a wrinkle-textured latex coating, with a aramid fiber liner for a high level of protection.

  • Reinforced Kevlar™ aramid fiber liner provides resistance against cuts and abrasion
  • Soft cotton lining wicks away sweat and moisture to ensure user comfort
  • Wrinkle-textured latex coating enhances grip capabilities
  • Ideal for use in airports and warehouses

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance EN 420:2003 + A1:2009, EN 388:3242B, ANSI/ISEA 105-2005, TAA Compliant
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Cut and Sewn
Coating Material Latex
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Aramid Fiber
Product Type Dip-coated Gloves
Size 10
EN388 Standard 3242B
For Use With (Application) Heavy Equipment Handling, HVAC, Public Utilities, and Window Manufacturing
Glove Cuff Color Natural
Glove Cuff Style Safety
Grip Design Crinkle Finish
Liner Material Cotton
Series 68NFW
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.