Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Triple-Layer Foam Insulated Cold-Protection Gloves

Catalog No. 19014967 Shop All Showa Best Glove Products
Change view
Click to view available options
Coated Area:
Full
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coated Area
19-014967 Full
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-014967 Supplier SHOWA™ Supplier No. 95NFW10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triple layer insulated gloves with an extended gauntlet cuff provide protection in extreme environments. An unique, self-wrinkle finish offers an enhanced grip in wet and dry conditions.

  • Triple-layered foam insulation for protection in cold environments
  • Self-wrinkle finish enhances grip in wet and dry conditions
  • Interlock cotton liner seals in warmth to keep cold out
  • Extended gauntlet cuff

Specifications

Coated Area Full
Coating Material Latex
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Size 10
For Use With (Application) Construction, Glass Handling, Landscaping, and Municipal Services
Glove Cuff Style Gauntlet
Liner Material Foam Insulation
Series 95NFW
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.