Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Heavy Duty PVC Coated Seamless Knit Cotton Gloves

Catalog No. 19014500 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-014-500 Small
11-395-16C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-014-500 Supplier SHOWA™ Supplier No. 960S08
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PVC-coating minimizes the absorption of water, oil, and grease with a rough-texture to enhance grip capabilities.

  • PVC-coating ensures breathability
  • Easy donning and doffing with slip-on cuff
  • Minimizes absorption of water, oil, and grease
  • Textured to enhance grip capabilities

Specifications

Chemotherapeutic Rated No
Coated Area Full Glove
Construction Seamless Knit
Coating Material PVC
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Material PVC
Product Type Seamless Knit Gloves
Size Small
Style Slip-on
For Use With (Application) Chemical Spray/Treatment, Utility Work, Intricate Parts Handling, and Oil-Coated Pieces
Gender Unisex
Glove Cuff Style Continuous Knit
Grip Design Rough
Liner Material Cotton
Packaging Type Polybag
Series 960
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.