Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 961 PVC Coated Seamless Cotton Lined Gloves

Catalog No. 19014517 Shop All Showa Best Glove Products
Change view
Click to view available options
Coating Material:
PVC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating Material
19-014517 PVC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-014517 Supplier SHOWA™ Supplier No. 96108
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a comfortable seamless cotton liner, with a fully textured PVC coating for enhanced grip capabilities.

  • Seamless cotton liner
  • Fully textured PVC coating
  • Knit wrist

Specifications

Coated Area Full
Construction Seamless Knit
Applications Industrial
Coating Material PVC
Color Yellow
Disposable Reusable
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Product Type General Purpose Gloves
Size 8
Sterility Non-sterile
For Use With (Application) Woodworking, assembly, shipping, and receiving
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Liner Material Cotton
Lining Knit Lining
Packaging Type Polybag
Palm Type Standard
Quantity 12 Pairs/Polybag, 72 Pairs/Cs.
Shell Material Cotton
Product Line SHOWA 961
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.