Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Neo Hyde™ Gauntlets

Catalog No. 19019281 Shop All Showa Best Glove Products
Change view
Click to view available options
Neo Hyde Gauntlet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-019281 Neo Hyde Gauntlet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-019281 Supplier SHOWA™ Supplier No. 36510
This item is not returnable. View return policy

Provides abrasion resistance as well as protection from oil, acids and caustics

Unique, etched-on, wrinkle-grip finish promotes an excellent grip, wet or dry, for safer, easier handling as well as additional protection from rough surfaces like metal castings and steel stampings.

  • Soft, cotton jersey liner
  • Fully coated, heavy-duty PVC
  • Sanitized™
  • Length: 12 in. (30.5 cm)
  • Launderable

Specifications

Certifications/Compliance (Europe) CE
Color Black
Material Vinyl
Size Large
For Use With (Application) Manufacturing, heavy duty maintenance, handling equipment with acids, caustics, greases and oil
Glove Cuff Style Gauntlet
Liner Material Cotton Jersey
Lining Lined
Model 365-10
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.