Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique BestHealth™ Sore Throat Lozenges

Catalog No. 19090770 Shop All Medique Products
Change view
Click to view available options
Quantity:
18/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-770 18/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-770 Supplier Medique Supplier No. 17818

For fast-acting relief from sore throat and minor mouth pain.

  • Each lozenge contains Menthol 3.6mg and Benzocaine 15mg

Specifications

For Use With (Application) Fast-Acting Relief From Sore Throat And Minor Mouth Pain
Product Type Sore Throat Lozenges
Format Menthol 3.6 mg and Benzocaine 15 mg
Quantity 18/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.