Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BET1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588961 Supplier Invitrogen™ Supplier No. PA588961
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRRAGLGEGV PPGNYGNYGY ANSGYSACEE ENERLTESLR SKVTAIKSLS IEIGHEVKTQ NKLLAEMDSQ FDSTTGFLGK TMGKLKILSR GSQT; Positive Samples: U-87MG, HeLa; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus, Golgi apparatus membrane, Single-pass type IV membrane protein, cis-Golgi network membrane.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BET1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BET1
Gene Accession No. O15155
Gene Alias BET1; Bet1 golgi vesicular membrane trafficking protein; BET1 homolog; Bet1p homolog; blocked early in transport 1 homolog; Golgi vesicular membrane trafficking protein p18; golgi vesicular membrane-trafficking protein p18; hBET1; rBET1
Gene Symbols BET1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-94 of human BET1 (NP_0058591).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10282
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.