Learn More
QA Bio Inc Beta-(1-3,4,6)-Galactosidase - 60 µls
Supplier:  QA Bio Inc EBG02
Encompass_Preferred
Beta-(1-3,4,6) Galactosidase cleaves all β(1-3) and β(1-4) linked non-reducing, terminal galactose. β(1-6) linked galactose is released at a slower rate. The enzyme is a glycoprotein. • β(1-4) galactose is by far the most common linkage found in N-linked oligosaccharides. For cleavage of this linkage we recommend β(1-4) Galactasidase (E-BG07). • Source: Bovine Testes • Contents: 200 µls of Beta-(1-3,4,6) Galactosidase in 20 mM Tris-HCl, 50 mM NaCl, 0.5mg/ml BSA,pH 7.5 / 5x Reaction Buffer- 500 mM sodium citrate/phosphate pH 4 • Specific Activity >10.2 U/mg • Activity >2.5 U/ml • Molecular weight 68,000 daltons • Specifictity Cleaves all ß(1-3) and ß(1-4) linked non-reducing, terminal galactose.
Catalog No. 50-223-6635
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.