Learn More
BioVendor, LLC Beta 2-Microglobulin Hs Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL014R
Encompass
Urinary excretion of Beta 2-microglobulin (beta 2M) is a widely used test of renal proximal tubular function. Proximal tubular dysfunction leads to an increased urinary concentration. The serum level of beta 2-microglobulin is determined by the glomerular filtration rate and the rate of synthesis. Increased production, with raised serum levels, is sometimes observed in malignancy, mainly at an advanced state. A strong correlation between lupus nephritis (LN), disease activity, and serum beta 2-microglobulin (b2MG) was observed. Serum b2MG level can be used as a valuable predictor for LN, clinical disease activity, and damage score. One kit (96 wells). Type: sandwich ELISA. Sample Type: serum, plasma, urine. Sample requirement 1 ¿l/well. Calibration range: 1.25-40ng/ml. Limit of detection: 0.4ng/ml.
Catalog No. 50-240-6379
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.