Learn More
Invitrogen™ beta-3 Adrenergic Receptor Polyclonal Antibody

Description
Immunogen sequence: MAPWPHENSS LAPWPDLPTL APNTANTSGL PGVPWEAALA GALLALAVLA TVGGNLLVIV AIAWTPRLQT MTNVFVTSLA AADLVMGLLV VPPAATLALT; Positive Samples: HL-60, K-562; Cellular Location: Cell membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | beta-3 Adrenergic Receptor |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ADRB3 |
| Gene Accession No. | P13945, P25962, P26255 |
| Gene Alias | ADRB; ADRB3; Adrb-3; Adrb3r; adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; adrenoceptor beta 3; B3AR; B3bar; beta 3 adrenoceptor; beta 3-AR; beta-3 adrenergic receptor; Beta-3 adrenoceptor; beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor; BETA3AR; Beta3-AR; FLJ99960 |
| Gene Symbols | ADRB3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.