Learn More
Invitrogen™ beta-5 Defensin Polyclonal Antibody, PeproTech®, Invitrogen™
Description
AA Sequence of recombinant protein: GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-5. Anti-Human BD-5-specific antibody was purified by affinity chromatography employing an immobilized Human BD-5 matrix. Sandwich ELISA: To detect Human BD-5 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-5 (500-P323Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-5. Western Blot: To detect hBD-5 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBD-5 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Specifications
Specifications
| Antigen | beta-5 Defensin |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | DEFB105A |
| Gene Accession No. | Q8NG35 |
| Gene Symbols | DEFB105A, DEFB105B |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.